6-Bromopiperonal 98%

Supplier: Origen Biomedical, Inc
Warning

A17510-14 A17510-06
AAA17510-14EA 232 USD
AAA17510-14 AAA17510-06
6-Bromopiperonal 98%
6-Bromopiperonal

Purity: 98% CAS No.: 15930-53-7 Molecular Formula: C8H5BrO3 Formula Weight: 229.03 Melting Point: 127-130° MDL No.: MFCD00022952

Formula: C₈H₅BrO₃
MW: 229.03 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00022952
CAS Number: 15930-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR