Triphenylsilanol 98%

Supplier: Origen Biomedical, Inc
Warning

A19284-14 A19284-22 A19284-06
AAA19284-14EA 81 USD
AAA19284-14 AAA19284-22 AAAA19284-06
Triphenylsilanol 98%
Triphenylsilanol

Purity: 98% CAS No.: 791-31-1 Molecular Formula: C18H16OSi Formula Weight: 276.41 Melting Point: 149-154° Beilstein Reference: 985007

Formula: C₁₈H₁₆OSi
MW: 276.41 g/mol
Melting Pt: 149…154 °C
Density: 1.17
Storage Temperature: Ambient
MDL Number: MFCD00002102
CAS Number: 791-31-1
EINECS: 212-339-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR