Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hexabromoplatinate ≥99%

Warning

78-1950-1G 78-1950-5G
100205-206EA 217.97 USD InStock
100205-206 100202-428
Potassium hexabromoplatinate ≥99%
Potassium hexabromoplatinate


Formula: Br₆K₂Pt
Melting Pt: 400 °C (dec.)
Density: 4,66
MDL Number: MFCD00011377
CAS Number: 16920-93-7
EINECS: 240-977-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty