Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium tetrafluoroborate

Danger

93-1951-500G 93-1951-2000G
100200-526EA 99.08 USD InStock
100200-526 100200-062
Potassium tetrafluoroborate
Potassium tetrafluoroborate


Formula: BF₄K
Boiling Pt: 960.85 °C (310 -rr)
Melting Pt: 96.5 - 98 °C
Density: 3.348 g/cm³ (25 °C)
MDL Number: MFCD00011395
CAS Number: 14075-53-7
EINECS: 237-928-2
UN: 1759
ADR: 8,II
Merck Index: 12,07859
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty