Copper(II) carbonate basic
|
Warning
|
93-2911-500G
93-2911-2000G
100209-076EA
90.58
USD
100209-076
100197-796
Copper(II) carbonate basic
Copper(II) carbonate basic
Formula:
CuCO₃·Cu(OH)₂ MW: 221.12 g/mol Melting Pt: 200 (dec.) Density: 4.0 |
MDL Number:
MFCD00010976 CAS Number: 12069-69-1 EINECS: 235-113-6 UN: 3288 ADR: 6.1 III Merck Index: 13,02658 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...