Cesium sulfate ≥99.9% (Cs basis)

93-5547-50G 93-5547-10G
100205-658EA 182.58 USD
100205-658 100204-366
Cesium sulfate ≥99.9% (Cs basis)
Cesium sulphate
Formula: Cs₂O₄S
MW: 361.87 g/mol
Boiling Pt: 1627
Melting Pt: 1010
Density: 4.243
Storage Temperature: Ambient
MDL Number: MFCD00010959
CAS Number: 10294-54-9
EINECS: 233-662-6
Merck Index: 13,02025

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR