Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyric acid ≥98%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

B1195-20LTBL B1195-200LTB
700004-780EA 998.07 USD InStock
700004-780 700004-778
Butyric acid ≥98%, Reagent Grade for synthesis
Butyric acid


Formula: C₄H₈O₂
Boiling Pt: 163.5 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.958 g/cm³ (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8,III
Merck Index: 13,01593
Order Now
Specification Test Results
Assay 98% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty