Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron(III) chloride hexahydrate ≥97%, lumps ACS suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

F1010-12KG F1010-50KGBL F1010-125GM F1010-2.5KG F1010-500GM F1010-25GM
700001-088EA 5157.75 USD InStock
700001-088 700001-096 700001-086 700001-090 700001-094 700001-092
Iron(III) chloride hexahydrate ≥97%, lumps ACS suitable for use as excipient
Iron(III) chloride hexahydrate
Ferric Chloride, Hexahydrate, Lump, Reagent, ACS, also known as Iron Chloride, is a chemical compound to produce the Weigert’s iron hematoxylin solution for nuclear stains, trichrome staining and staining with acidic counterstains As an ACS grade Reagent, Spectrum Chemical manufactured Ferric Chloride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity


Formula: FeCl₃·6H₂O
Boiling Pt: 280 - 285 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=fd3b4ce92b750af3127aba38a6faded5be69de4a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 97% Min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty