Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

P1216-100GM P1216-500GM
95035-240EA 203.9 USD
95035-240 95035-242
Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 FCC suitable for use as excipient
Polyvinylpyrrolidone

Polyvinylpyrrolidone K-30, FCC is used as an iodine antiseptic. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR