1,3,5,7-Cyclooctatetraene 98% stabilised
Supplier: Origen Biomedical, Inc
|
Danger
|
Stabilization:
0.1% Stabilised with hydroquinone (HQ)
B22636-06
B22636-14
AAAB22636-06EA
182
USD
AAAB22636-06
AAB22636-14
1,3,5,7-Cyclooctatetraene 98% stabilised
1,3,5,7-Cyclooctatetraene
Purity: 98%
CAS No.: 629-20-9
Molecular Formula: C8H8
Formula Weight: 104.15
Storage Temperature: 2°C to 8°C
Flash Point: 20°(68°F)
Refractive Index: 1.537
Odor: Unpleasant
Melting Point: -5°
Boiling Point: 142-143°
Beilstein Reference: 2496800
UN No.: 2358
MDL No.: MFCD00004161
Formula:
C₈H₈ MW: 104.15 g/mol Boiling Pt: 143 Melting Pt: -7 Density: 0.925 Flash Pt: 20 °C (68 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004161 CAS Number: 629-20-9 EINECS: 211-080-3 UN: 2358 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...