Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,5-Dimethoxyphenyl)acetic acid 99%

Supplier: Origen Biomedical, Inc
Warning

H27328-06 H27328-14
AAH27328-06EA 63.7 USD InStock
AAH27328-06 AAH27328-14
(2,5-Dimethoxyphenyl)acetic acid 99%
(2,5-Dimethoxyphenyl)acetic acid
Purity: 99% CAS No.: 1758-25-4 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 123-125° MDL No.: MFCD00004322


Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Boiling Pt: 327
Melting Pt: 123
Storage Temperature: Ambient
MDL Number: MFCD00004322
CAS Number: 1758-25-4
EINECS: 217-151-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty