2-Fluorobenzophenone 98+%

Supplier: Origen Biomedical, Inc
Warning

L09316-14 L09316-06
AAAL09316-14EA 115 USD
AAAL09316-14 AAAL09316-06
2-Fluorobenzophenone 98+%
2-Fluorobenzophenone

Purity: 98+% CAS No.: 342-24-5 Molecular Formula: C13H9FO Formula Weight: 200.21 Flash Point: 110°(230°F) Refractive Index: 1.5864 Boiling Point: 149-150°/16mm Beilstein Reference: 2047045 MDL No.: MFCD00000318

Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 149…150 °C (16 mmHg)
Density: 1.186
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000318
CAS Number: 342-24-5
EINECS: 206-440-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR