VWR® Student Grade Filter Paper
Supplier: GE Healthcare - Whatman
28310-015 28310-026 28310-048 28310-081 28310-106 28310-128 28310-161 28310-172 28310-208
470204-452EA 9.2 USD
470204-452 470204-454 470204-456 470204-458 470204-460 470204-462 470204-464 470204-466 470204-468
VWR® Student Grade Filter Paper
Filters Filter Paper Qualitative Filter Paper
For student requirements in studying general chemistry, these qualitative papers are designed for filtration of aluminum phosphate, barium carbonate, lead chromate, and more.
  • Porosity: Medium
  • Particle Retention: 5 μm
  • Flow Rate: Medium
Envase: Package of 100.