Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-chloro-4-fluorobenzene ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

001614-25G 001614-100G
101782-090EA 23.08 USD InStock
101782-090 101782-092
1-Bromo-3-chloro-4-fluorobenzene ≥98% for synthesis
1-Bromo-3-chloro-4-fluorobenzene
Hazard code(s): Irritant


Formula: C₆H₃BrClF
Boiling Pt: 92 °C (35 -rr)
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00051794
CAS Number: 60811-21-4
EINECS: 405-580-2
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Boiling point 194°C
Density 1.727
ND20 1.553

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty