Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,6,6-Tetramethyl-3,5-heptanedione ≥95%

Supplier: MATRIX SCIENTIFIC MS

003516-25G 003516-100G
101785-824EA 58.46 USD InStock
101785-824 101785-826
2,2,6,6-Tetramethyl-3,5-heptanedione ≥95%
2,2,6,6-Tetramethyl-3,5-heptanedione
Hazard code(s): Irritant


Formula: C₁₁H₂₀O₂
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 28 - 30 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4
EINECS: 214-268-3
Order Now
Specification Test Results
Boiling point 72-73°C/6mm
Density 0.883
ND20 1.459

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty