Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-4-nitro-o-toluidine ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Danger

045961-1G 045961-5G
101873-474EA 46.15 USD InStock
101873-474 101873-476
6-Chloro-4-nitro-o-toluidine ≥95% for synthesis
6-Chloro-4-nitro-o-toluidine
Hazard code(s): Irritant


Formula: C₇H₇ClN₂O₂
Melting Pt: 172.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01313703
CAS Number: 69951-02-6
UN: 2237
ADR: 6.1,III
Order Now
Specification Test Results
Melting point 165-167°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty