Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-fluoro-6-nitrobenzene ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

054377-1G 054377-5G 054377-25G
101907-192EA 41.54 USD InStock
101907-192 101907-194 101907-196
1-Bromo-2-fluoro-6-nitrobenzene ≥98% for synthesis
1-Bromo-2-fluoro-6-nitrobenzene
Hazard code(s): Irritant


Formula: C₆H₃BrFNO₂
Melting Pt: 42…45 °C
MDL Number: MFCD07782080
CAS Number: 59255-94-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty