Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(4-methylbenzyloxy)benzonitrile

Supplier: MATRIX SCIENTIFIC MS

002732-1G 002732-5G
101784-538EA 36.92 USD InStock
101784-538 101784-540
2-Fluoro-6-(4-methylbenzyloxy)benzonitrile
2-Fluoro-6-(4-methylbenzyloxy)benzonitrile
Hazard: Irritant


Formula: C₁₅H₁₂FNO
Melting Pt: 76…78 °C
MDL Number: MFCD00068204
CAS Number: 175204-09-8
Order Now
Specification Test Results
Melting point 76-78°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty