Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Chloro-2-phenyl-4-pyrimidinyl)methylmethyl ether ≥95%

Supplier: MATRIX SCIENTIFIC MS

044501-500MG 044501-1G 044501-5G
101866-200EA 192.31 USD InStock
101866-200 101866-202 101866-204
(6-Chloro-2-phenyl-4-pyrimidinyl)methylmethyl ether ≥95%
(6-Chloro-2-phenyl-4-pyrimidinyl)methylmethyl ether
Hazard: Irritant


Formula: C₁₂H₁₁ClN₂O
Melting Pt: 54…56 °C
MDL Number: MFCD00794769
CAS Number: 325685-59-4
Order Now
Specification Test Results
Melting point 54-56°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty