Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Pyrazol-3-yl)-1,3-thiazole ≥95%

Supplier: MATRIX SCIENTIFIC MS

044970-500MG 044970-1G 044970-5G
101868-502EA 307.69 USD InStock
101868-502 101868-504 101868-506
2-(1H-Pyrazol-3-yl)-1,3-thiazole ≥95%
2-(1H-Pyrazol-3-yl)-1,3-thiazole
Hazard: Irritant


Formula: C₆H₅N₃S
Melting Pt: 154…156 °C
MDL Number: MFCD00105072
CAS Number: 166196-73-2
Order Now
Specification Test Results
Melting point 154-156°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty