Ethyl-1-(2,3-diethyl-6-quinoxalinyl)-4-piperidinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045123-500MG 045123-1G
101869-248EA 556.92 USD
101869-248 101869-250
Ethyl-1-(2,3-diethyl-6-quinoxalinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(2,3-diethyl-6-quinoxalinyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₂₀H₂₇N₃O₂
MW: 341.46 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD03425796
CAS Number: 439095-56-4

Order Now

Specification Test Results

Melting point 86-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR