Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulfonate ≥97%

Supplier: MATRIX SCIENTIFIC MS

004886-5G 004886-25G 004886-100G
101787-956EA 41.54 USD InStock
101787-956 101787-958 101787-960
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulfonate ≥97%
1,1,1,3,3,3-Hexafluoroisopropyl toluene-4-sulphonate
TCA listed: T
Hazard: Irritant


Formula: C₁₀H₈F₆O₃S
Melting Pt: 39-40°C
MDL Number: MFCD00039262
CAS Number: 67674-48-0
Order Now
Specification Test Results
Melting point 39-40°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty