3,4-Pyridinedicarboxylic acid anhydride ≥97%

Supplier: MATRIX SCIENTIFIC MS

021397-5G 021397-25G
101811-966EA 75.38 USD
101811-966 101811-968
3,4-Pyridinedicarboxylic acid anhydride ≥97%
3,4-Pyridinedicarboxylic acid anhydride

Hazard: Irritant

Formula: C₇H₃NO₃
MW: 149.11 g/mol
Boiling Pt: 139…142 °C (12 Torr)
Melting Pt: 73 °C
MDL Number: MFCD00010680
CAS Number: 4664-08-8

Order Now

Specification Test Results

Melting point 75-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR