N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-6-carboxamide ≥90%
Supplier: MATRIX SCIENTIFIC MS
046872-500MG
046872-1G
101877-386EA
921.54
USD
101877-386
101877-388
N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-6-carboxamide ≥90%
N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-6-carboxamide
Hazard: Irritant
Formula:
C₁₀H₁₁N₃O₂ MW: 205.22 g/mol Melting Pt: 200…202 °C |
MDL Number:
MFCD09027091 CAS Number: 937604-34-7 |
Specification Test Results
Melting point | 200-202°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...