Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-{[(4-methoxyphenyl)methyl]amino}-3-nitrobenzoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

059662-500MG 059662-1G
101918-816EA 315.38 USD InStock
101918-816 101918-818
Methyl-4-{[(4-methoxyphenyl)methyl]amino}-3-nitrobenzoate ≥95%
Methyl-4-{[(4-methoxyphenyl)methyl]amino}-3-nitrobenzoate
Hazard: Irritant


Formula: C₁₆H₁₆N₂O₅
Melting Pt: 159…160 °C
MDL Number: MFCD18803533
CAS Number: 234751-03-2
Order Now
Specification Test Results
Melting point 159-160°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty