4-(4-Fluorobenzoyl)butyric acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

007076-25G 007076-100G
101791-968EA 21.54 USD
101791-968 101791-970
4-(4-Fluorobenzoyl)butyric acid ≥97%
4-(4-Fluorobenzoyl)butyric acid

Purity: 97%

Formula: C₁₁H₁₁FO₃
MW: 210.20 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD00667222
CAS Number: 149437-76-3

Order Now

Specification Test Results

Melting point 142-144°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR