tert-Butyl-4-aminophenylacetate ≥98%

Supplier: MATRIX SCIENTIFIC MS

012544-1G 012544-5G
101802-810EA 152.31 USD
101802-810 101802-812
tert-Butyl-4-aminophenylacetate ≥98%
tert-Butyl-4-aminophenylacetate

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Melting Pt: 30…32 °C
MDL Number: MFCD06245494
CAS Number: 174579-31-8