Tridecafluoroheptanamide ≥97%

Supplier: MATRIX SCIENTIFIC MS

012892-1G 012892-5G
101803-572EA 80 USD
101803-572 101803-574
Tridecafluoroheptanamide ≥97%
Tridecafluoroheptanamide

Hazard: Irritant

Formula: C₇H₂F₁₃NO
MW: 363.08 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00407510
CAS Number: 2358-22-7

Order Now

Specification Test Results

Melting point 136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR