Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,5-dimethylbenzenesulfonamide ≥98%

Supplier: MATRIX SCIENTIFIC MS

020746-1G 020746-5G
101810-692EA 83.08 USD InStock
101810-692 101810-694
4-Chloro-2,5-dimethylbenzenesulfonamide ≥98%
4-Chloro-2,5-dimethylbenzenesulfonamide
Hazard: Irritant


Formula: C₈H₁₀ClNO₂S
Melting Pt: 190…192 °C
MDL Number: MFCD00275263
CAS Number: 219689-73-3
Order Now
Specification Test Results
Melting point 190-192°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty