Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl 4-ethylbenzenesulfonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042813-500MG 042813-1G 042813-5G
101857-674EA 375.38 USD InStock
101857-674 101857-676 101857-678
Pentafluorophenyl 4-ethylbenzenesulfonate ≥95%
Pentafluorophenyl 4-ethylbenzenesulphonate
Hazard: Irritant


Formula: C₁₄H₉F₅O₃S
Melting Pt: 61-63°C
MDL Number: MFCD05975134
CAS Number: 885950-41-4
Order Now
Specification Test Results
Melting point 61-63°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty