Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(2,5-Dimethylbenzyl)sulfanyl]benzene carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

043058-500MG 043058-5G
101858-934EA 375.38 USD InStock
101858-934 101858-938
2-[(2,5-Dimethylbenzyl)sulfanyl]benzene carboxylic acid ≥95%
2-[(2,5-Dimethylbenzyl)sulfanyl]benzene carboxylic acid
Hazard: Irritant


Formula: C₁₆H₁₆O₂S
Melting Pt: 193…196 °C
MDL Number: MFCD03001236
CAS Number: 385383-43-7
Order Now
Specification Test Results
Melting point 193-196°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty