Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-chloro-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

043245-500MG 043245-1G 043245-5G
101859-862EA 375.38 USD InStock
101859-862 101859-864 101859-866
Methyl-5-chloro-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-5-chloro-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate
Hazard: Irritant


Formula: C₁₂H₉ClN₂O₃
MW: 264.67 g/mol
Melting Pt: 123…126 °C
MDL Number: MFCD01109226
CAS Number: 401571-11-7
Order Now
Specification Test Results
Melting point 123-126°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty