Imidazo[1,2-a]pyridin-8-ol ≥95%
Supplier: MATRIX SCIENTIFIC MS
045760-500MG
045760-1G
101872-450EA
184.62
USD
101872-450
101872-452
Imidazo[1,2-a]pyridin-8-ol ≥95%
Imidazo[1,2-a]pyridin-8-ol
Hazard: Irritant
Formula:
C₇H₆N₂O MW: 134.1400 g/mol Melting Pt: 154…157 °C |
MDL Number:
MFCD06496222 CAS Number: 69214-22-8 |
Specification Test Results
Melting point | 154-157°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...