Ethyl-1-(6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate ≥95%
Supplier: MATRIX SCIENTIFIC MS
047810-500MG
047810-1G
101881-542EA
246.15
USD
101881-542
101881-544
Ethyl-1-(6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₇N₃O₃ MW: 251.29 g/mol Melting Pt: 187…188 °C |
MDL Number:
MFCD09864836 CAS Number: 1000018-23-4 |
Specification Test Results
Melting point | 187-188°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...