Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Allyl-3-(2-hydroxyethyl)-2-thiourea

Supplier: MATRIX SCIENTIFIC MS

056880-50G 056880-250G
101911-580EA 201.54 USD InStock
101911-580 101911-582
1-Allyl-3-(2-hydroxyethyl)-2-thiourea
1-Allyl-3-(2-hydroxyethyl)-2-thiourea
Hazard: Irritant


Formula: C₆H₁₂N₂OS
Melting Pt: 77…79 °C
MDL Number: MFCD00002838
CAS Number: 105-81-7
Order Now
Specification Test Results
Melting point 77-79°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty