Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)-3-ethoxydodecafluorohexane ≥99%

Supplier: MATRIX SCIENTIFIC MS

009203-25G 009203-100G
101796-632EA 180 USD InStock
101796-632 101796-634
2-(Trifluoromethyl)-3-ethoxydodecafluorohexane ≥99%
2-(Trifluoromethyl)-3-ethoxydodecafluorohexane
TCA listed: T
Hazard: Irritant


Formula: C₉H₅F₁₅O
Boiling Pt: 130 °C
Melting Pt: –100 °C
Density: 1.61
MDL Number: MFCD04038367
CAS Number: 297730-93-9
Order Now
Specification Test Results
Boiling point 130°C
Melting point -100°C
Density 1.61

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty