Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Trichloroethyl acrylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

009177-1G 009177-5G
101796-572EA 53.85 USD InStock
101796-572 101796-574
2,2,2-Trichloroethyl acrylate ≥97%
2,2,2-Trichloroethyl acrylate
Hazard: Irritant/Refrigerate


Formula: C₅H₅Cl₃O₂
Boiling Pt: 31.5 °C (0.5 mmHg)
Density: 1.376
MDL Number: MFCD04038361
CAS Number: 44925-09-9
Order Now
Specification Test Results
Boiling point 31.5°C/0.5m
Density 1.376
ND20 1.4738

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty