Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Perfluorooctyl)prop-2-enol ≥98%

Supplier: MATRIX SCIENTIFIC MS

007118-5G 007118-25G
101792-062EA 144.62 USD InStock
101792-062 101792-064
3-(Perfluorooctyl)prop-2-enol ≥98%
3-(Perfluorooctyl)prop-2-enol
Hazard: Irritant


Formula: C₁₁H₅F₁₇O
Boiling Pt: 82-85°C/8mm
Density: 1.718
MDL Number: MFCD00042361
CAS Number: 2340-84-3
Order Now
Specification Test Results
Boiling point 82-85°C/8mm
Density 1.718
ND20 1.33

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty