2-Bromo-5-(trifluoromethyl)phenol ≥98%

Supplier: MATRIX SCIENTIFIC MS

059835-1G 059835-5G
101919-230EA 30.77 USD
101919-230 101919-232
2-Bromo-5-(trifluoromethyl)phenol ≥98%
2-Bromo-5-(trifluoromethyl)phenol

Hazard: Irritant

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Boiling Pt: 219 °C
Density: 1.752
Flash Pt: 86.3 °C
MDL Number: MFCD09261190