Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole

Supplier: MATRIX SCIENTIFIC MS

006181-1G 006181-5G
101790-208EA 27.69 USD InStock
101790-208 101790-210
1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Hazard: Irritant


Formula: C₁₄H₁₁F₆N
Boiling Pt: 78…80 °C (0.1 mmHg)
MDL Number: MFCD00084902
CAS Number: 175205-51-3
Order Now
Specification Test Results
Boiling point 78-80°C/0.1

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty