Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Dichloro-2-iodo-1,2,2-trifluoroethane ≥70%

Supplier: MATRIX SCIENTIFIC MS

004996-5G 004996-25G 004996-100G
101788-122EA 27.69 USD InStock
101788-122 101788-124 101788-126
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane ≥70%
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane
Balance is 1,2-Dichloro-1-iodotrifluoroethane

Hazard: Irritant


Formula: C₂Cl₂F₃I
Boiling Pt: 53 °C (150 mmHg)
Density: 2.2
MDL Number: MFCD02093303
CAS Number: 661-66-5
Order Now
Specification Test Results
Density 2.2

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty