Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥98%

Supplier: MATRIX SCIENTIFIC MS

002650-5G 002650-25G
101784-392EA 30.77 USD InStock
101784-392 101784-394
2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥98%
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Hazard: Irritant


Formula: C₈H₆ClF₃O
Melting Pt: 57-60°C
MDL Number: MFCD01631469
CAS Number: 64372-62-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty