2-Oxo-3-piperidinecarboxylic acid

Supplier: MATRIX SCIENTIFIC MS

053737-500MG 053737-2.5G
101904-814EA 538.46 USD
101904-814 101904-816
2-Oxo-3-piperidinecarboxylic acid
2-Oxo-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₆H₉NO₃
MW: 143.14 g/mol
MDL Number: MFCD08061931
CAS Number: 41888-21-5