Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Morpholinium hexafluorophosphate ≥97%

Supplier: MATRIX SCIENTIFIC MS

004248-5G 004248-25G
101787-016EA 144.62 USD InStock
101787-016 101787-018
Morpholinium hexafluorophosphate ≥97%
Morpholinium hexafluorophosphate
Hazard: Irritant


Formula: C₄H₁₀F₆NOP
MDL Number: MFCD00067328
CAS Number: 25866-83-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty