Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,3S)-Fmoc-3-aminocyclopentane-1-carboxylic acid

Supplier: MATRIX SCIENTIFIC MS

041319-250MG 041319-1G
101851-326EA 300 USD InStock
101851-326 101851-328
(1R,3S)-Fmoc-3-aminocyclopentane-1-carboxylic acid
(1R,3S)-Fmoc-3-aminocyclopentane-1-carboxylic acid
Hazard: Irritant


Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
MDL Number: MFCD01311758
CAS Number: 220497-67-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty