Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Tic-OH

Supplier: MATRIX SCIENTIFIC MS

041916-1G 041916-5G
101853-718EA 101.54 USD InStock
101853-718 101853-720
Fmoc-Tic-OH
Fmoc-Tic-OH
Hazard: Irritant


Formula: C₂₅H₂₁NO₄
MDL Number: MFCD00144368
CAS Number: 136030-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty