2-Ethynyl-4-methylaniline 95

Supplier: MATRIX SCIENTIFIC MS

054238-500MG 054238-1G
101906-714EA 461.54 USD
101906-714 101906-716
2-Ethynyl-4-methylaniline 95
2-Ethynyl-4-methylaniline

Hazard: Irritant

Formula: C₉H₉N
MW: 131.18 g/mol
MDL Number: MFCD11036439
CAS Number: 215589-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR