5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde ≥95%
Supplier: MATRIX SCIENTIFIC MS
054245-500MG
054245-1G
101906-746EA
706.15
USD
101906-746
101906-748
5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde ≥95%
5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde
Hazard: Irritant
Formula:
C₈H₅FN₂O MW: 164.14 g/mol |
MDL Number:
MFCD11656369 CAS Number: 878197-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...