Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-1-Ethoxyethene-2-boronic acid pinacol ester ≥97%

Supplier: MATRIX SCIENTIFIC MS

069626-1G 069626-5G 069626-25G
101941-150EA 230.77 USD InStock
101941-150 101941-152 101941-154
(Z)-1-Ethoxyethene-2-boronic acid pinacol ester ≥97%
(Z)-1-Ethoxyethene-2-boronic acid pinacol ester
Hazard: Irritant/Store in Freezer


Formula: C₁₀H₁₉BO₃
MDL Number: MFCD09702441
CAS Number: 219489-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty