2-Amino-5-bromopyrazine ≥97%

Supplier: MATRIX SCIENTIFIC MS

069642-25G 069642-100G
101941-232EA 73.85 USD
101941-232 102551-754
2-Amino-5-bromopyrazine ≥97%
2-Amino-5-bromopyrazine

Hazard: Irritant

Formula: C₄H₄BrN₃
MW: 174.00 g/mol
Melting Pt: 113…117 °C
MDL Number: MFCD00235015
CAS Number: 59489-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR